Anti-CK14 Mouse Monoclonal Antibody [clone: LL002]

Supplier: Diagnostic BioSystems

MOB186-05 MOB186-05 MOB186 MOB186
95024-472EA 351.51 USD
CA95024-472 CA99990-392 95024-472 99990-392
Anti-CK14 Mouse Monoclonal Antibody [clone: LL002]
Antibodies
This antibody is specific to 50 kDa cytokeratin protein designated cytokeratin 14. Cytokeratin 14 belongs to the type A (acidic) subfamily of low molecular weight cytokeratins and exists in combination with cytokeratin 15. This antibody can be used to distinguish stratified epithelial cells from simple epithelial cells.

Type:
Antigen: CK14
Clonality: Monoclonal
Clone: LL002
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR