Anti-Cytokeratin Wide Spectrum Rabbit Polyclonal Antibody

Supplier: Diagnostic Biosystems

RP010 RP010-05 PDR035
99991-216EA 490.08 USD
99991-216 95024-990 99991-110
Anti-Cytokeratin Wide Spectrum Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with cytokeratins of 58, 56, 52, 60, 51, 48 and 68 kDa MW. This antibody is well suited for the staining of a broad spectrum of human keratins.

Type:
Antigen:
Clonality:
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR