Anti-Cytokeratin (Acidic/Basic) Mouse Monoclonal Antibody [clone: AE1 + AE3]

Supplier: Diagnostic Biosystems

PDM072 MOB190-05 MOB190
99990-928EA 320.24 USD
99990-928 95024-506 99990-398
Anti-Cytokeratin (Acidic/Basic) Mouse Monoclonal Antibody [clone: AE1 + AE3]
Antibodies
This AE1/AE3 antibody cocktail recognizes most of the acidic (Type I) keratins as well as basic (Type II). It recognizes keratin polypeptides of 40, 48, 50, 52, 54, 56.5, 58, 59 and 64 to 67 kDa. In keratinized epidermis, 50 kDa keratin is present in the basal layer, while 56.5 kDa keratin is present in suprabasal layers. 58 kDa keratin is present in the basal and suprabasal layers, while 65 to 67 kDa keratin is present in the cells above the basal layers. This antibody can be used to stain distinct cell layers in an epithelial tissue and various neoplasms of epithelial origin.

Type:
Antigen: Cytokeratin (Acidic/Basic)
Clonality: Monoclonal
Clone: AE1 + AE3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa + IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR