Anti-Cytokeratin (Acidic) Mouse Monoclonal Antibody [clone: AE1]

Supplier: Diagnostic Biosystems

PDM043 MOB092
99990-884EA 296.72 USD
99990-884 99990-284
Anti-Cytokeratin (Acidic) Mouse Monoclonal Antibody [clone: AE1]
Antibodies
This antibody recognizes most of the acidic (Type I) keratins. It recognizes keratin polypeptides of 50 and 56.5 kDa. In keratinized epidermis, 50 kDa keratin is present in the basal layer, while 56.5 kDa keratin is present in suprabasal layers. This antibody can be used to stain distinct cell layers in an epithelial tissue and various neoplasms of epithelial origin.

Type:
Antigen: Cytokeratin (Acidic)
Clonality: Monoclonal
Clone: AE1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR