Stage Micrometer Model SM-3, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60210-3PG 60210-3PGC
102093-112EA 714.16 USD
102093-112 102680-928
Stage Micrometer Model SM-3, Electron Microscopy Sciences
Micrometers
Linear Reference Scales are available in Chrome-on-Glass (G) and Chrome-on-Opal glass (O) or Chrome-on Quartz (Q)
Positive image are mostly clear or white.
Ordering information: The Opal and Quartz are offered as a special only and must be requested.
Packaging: All EMS micrometers come in a sturdy wooden box.