Hard Plastic Microscope Slide Tray, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71503-01 71503-10
102097-584EA 12.68 USD
102097-584 102097-586
Hard Plastic Microscope Slide Tray, Electron Microscopy Sciences
Slide Holders
This durable ABS hard plastic slide tray will hold up to 20 standard microscope slides 75 x 25 mm (3x1”).
Ivory color consists of two rows, each with securely holds 10 slides. Each numbered cavity has thumb cut and raise notches for easy to remove the slide. Stackable. Measures 11 7/8” L x 7 ”W x ½”H (300 x 195 x 12 mm).