Silver Conductive Adhesive Paste 478SS, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
12685-25
100502-398EA
18.35
USD
100502-398
100502-396
Silver Conductive Adhesive Paste 478SS, Electron Microscopy Sciences
Stains
Microscopy and staining reagents
Adhesive 478SS is a conductive, silver-based polymer which is used for thick film coatings where liquid silver is not an option.
- Conductive
- Very high Tg
- Superior adhesion to polyester film
Once cured, adhesive offers a very high Tg (glass transition temperature) 153°F (67°C) to prevent blocking, and offers superior adhesion to polyester film. Adhesive 478SS can be cured at 200°F (93°C) within 15 minutes. Higher temperatures will reduce the time needed to achieve a final cure.