Nitrobenzene ≥99% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.06770.0100 8.06770.1000 8.06770.2500 8.06770.9033
EM8.06770.0100CS 439 USD
EM8.06770.0100 EM8.06770.1000 EM8.06770.2500 EM8.06770.9033
Nitrobenzene ≥99% (by GC) for synthesis, Sigma-Aldrich®
Nitrobenzene
Formula: C₆H₅NO₂
MW: 123.11 g/mol
Boiling Pt: 210.8
Melting Pt: 5.7
Density: 1.20 g/cm3 (20 °C)
Flash Pt: 88
Storage Temperature: Ambient
MDL Number: MFCD00007043
CAS Number: 98-95-3
UN: 1662
ADR: 6.1 II
Merck Index: 13,06621

Order Now

Specification Test Results

Solubility 1.90 g/l (20 °C)
Melting Point 6.0 °C
Molar Mass 123.11 g/mol
Boiling Point 211 °C (1013 hPa)
Vapor Pressure 0.3 hPa (20 °C)
Flash Point 88 °C
Refractive Index 1.553 (20 °C, 589 nm)
Explosion Limit 1.8 - 40 %(V)
Density 1.20 g/cm3 (20 °C)
pH 8.1 (1 g/l, H2O, 20 °C)
Ignition Point 480 °C DIN 51794


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR