Type J Thermocouple Probe, Ace Glass Incorporated
Supplier: Ace Glass
12110-09 12110-15 12110-17 12110-24 12110-25 12110-26 12110-27
89076-446EA 178.73 USD
89076-446 89076-448 89076-450 89076-452 89076-454 89076-456 89076-458
Type J Thermocouple Probe, Ace Glass Incorporated
Electrodes pH Electrodes
Type J temperature probes provide economical measurement from -20° C to 550° C
Probes available sheathed with stainless steel, inconel or simply insulated wire. With male mini plug lead.