Triphenyl phosphate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Triphenylphosphate, TPP

M-Clarity™ quality level = MQ200

8.21197.0100 8.21197.1000
EM8.21197.0100CS 667 USD
EM8.21197.0100 EM8.21197.1000
Triphenyl phosphate for synthesis, Sigma-Aldrich®
Triphenyl phosphate
Formula: C₁₈H₁₅O₄P
MW: 326.29 g/mol
Boiling Pt: 412 °C (1013 hPa)
Melting Pt: 50…51 °C
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 223 °C
Storage Temperature: Ambient
MDL Number: MFCD00003031
CAS Number: 115-86-6
EINECS: 204-112-2
UN: 3077
ADR: 9,III
Merck Index: 12,09872

Order Now

Specification Test Results

Solubility 0.0019 g/l (25 °C)
Melting Point 48 - 50 °C
Molar Mass 326.28 g/mol
Bulk Density 650 kg/m3
Boiling Point 220 °C (5 hPa)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 230 °C
Ignition Point 500 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR