Cyanoguanidine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.02491.0100 8.02491.1000
EM8.02491.0100CS 473 USD
EM8.02491.0100 EM8.02491.1000
Cyanoguanidine for synthesis, Sigma-Aldrich®
Cyanoguanidine
Formula: (H₂N)₂CNCN
Boiling Pt: 143 °C (1013 hPa)
Density: 1.404 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008066
CAS Number: 461-58-5
EINECS: 207-312-8
Merck Index: 13,03119

Order Now

Specification Test Results

Solubility 34.6 g/l (20 °C)
Melting Point 208 - 210 °C
Molar Mass 84.08 g/mol
Bulk Density 400 kg/m3
Density 1.4 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR