Vincristine sulfate 98+%, powder
Supplier: Thermo Scientific Chemicals
|
Danger
|
J60907-MB
J60907-MA
AAJ60907-MBEA
355
USD
AAJ60907-MB
AAJ60907-MA
Vincristine sulfate 98+%, powder
Vincristine sulphate
An anti-tumor alkaloid used in cancer research
Formula:
C₄₆H₅₈N₄O₁₄S Boiling Pt: 273…281 °C Melting Pt: >300 °C Storage Temperature: Freezer |
MDL Number:
MFCD00084729 CAS Number: 2068-78-2 EINECS: 218-190-0 UN: 2811 ADR: 6.1,III Merck Index: 14,9986 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...