Bovine Serum Albumin (BSA)
Supplier: Rockland Immunochemical
BSA-1000
BSA-50
BSA-10
RLBSA-1000EA
1377.01
USD
RLBSA-1000
RLBSA50
RLBSA-10
Bovine Serum Albumin (BSA)
Proteins and Peptides
This product is suitable for use in protease sensitive assays such as RIA, EIA and nucleic acid hybridization, use as a stabilizing agent for proteins and enzymes including dilute solutions of antibody, use as a blocking agent to reduce non-specific binding.
- IgG-free
- Protease-free
This product is suitable for use in protease sensitive assays such as RIA, EIA and nucleic acid hybridization, use as a stabilizing agent for proteins and enzymes including dilute solutions of antibody, use as a blocking agent to reduce non-specific binding.