Foetal bovine serum, Australian origin, Corning®
Supplier: Corning
Corning®
35-086-CF 35-076-CV 35-076-CV
89414-566EA 1546.37 USD
89414-566 CA76332-844 CA89414-566
Foetal bovine serum, Australian origin, Corning®
Sera (Serum)
FBS is a commonly used cell culture media supplement which helps sustain cell growth and satisfy metabolic requirements.
  • Triple sterile filtered (0,1 μm)
  • Low endotoxin (<20 EU/ml)
  • Low haemoglobin (<30 mg/dl)
Corning® sera products are analysed for quality and performance. Testing includes pH, osmolality, growth promotion, sterility (current USP method), mycoplasma, total protein, endotoxin and haemogloblin levels, and viral contamination. A full biochemical profile is available for each lot upon request.