Hydroquinone for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
1,4-Dihydroxybenzene, Quinol
M-Clarity™ quality level = MQ200
8.22333.0250
8.22333.1000
8.22333.2501
EM8.22333.0250CS
863
USD
EM8.22333.0250
EM8.22333.1000
EM8.22333.2501
Hydroquinone for synthesis, Sigma-Aldrich®
Hydroquinone
Formula:
C₆H₄(OH)₂ MW: 110.11 g/mol Boiling Pt: 285 Melting Pt: 172 Density: 1.332 g/cm³ (20 °C) Flash Pt: 165 Storage Temperature: Ambient |
MDL Number:
MFCD00002339 CAS Number: 123-31-9 EINECS: 204-617-8 UN: 3077 ADR: 9 III Merck Index: 13,04833 |
|
Specification Test Results
Solubility | 70 g/l (25 °C) |
Melting Point | 172 °C (decomposition) |
Molar Mass | 110.11 g/mol |
Bulk Density | 600 kg/m3 |
Boiling Point | 287 °C (1013 hPa) |
Vapor Pressure |
|
Flash Point | 165 °C |
Density | 1.358 g/cm3 (20 °C) |
pH | (70 g/l, H2O, 20 °C) |
Ignition Point | 515 °C |
Hazard Class | 9 |
Hazard Class Description | Miscellaneous Hazardous Material |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...