Amidosulfonic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Aminosulphonic acid, Amidosulphuric acid, Sulfamic acid, Sulfamidic acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid, Aminosulfonic acid, Amidosulfonic acid, Amidosulfuric Acid, Amidosulfuric acid

M-Clarity™ quality level = MQ200

8.14483.0100 8.14483.1000
EM8.14483.0100CS 3325 USD
EM8.14483.0100 EM8.14483.1000
Amidosulfonic acid for synthesis, Sigma-Aldrich®
Amidosulphonic acid
Formula: H₃NO₃S
MW: 97.09 g/mol
Boiling Pt: 247
Melting Pt: 205
MDL Number: MFCD00011603
CAS Number: 5329-14-6
UN: 2967
ADR: 8 III
Merck Index: 13,09006
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (acidimetric) ≥ 99.0 %
Identity (IR) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR