Imidazole for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Glyoxaline, 1H-Imidazole, Glyoxalamine
M-Clarity™ quality level = MQ200
8.14223.0005
8.14223.0250
8.14223.1000
EM8.14223.0005CS
18505
USD
EM8.14223.0005
EM8.14223.0250
EM8.14223.1000
Imidazole for synthesis, Sigma-Aldrich®
Imidazole
Formula:
C₃H₄N₂ MW: 68.08 g/mol |
MDL Number:
MFCD00005183 CAS Number: 288-32-4 UN: 3263 ADR: 8,III |
Specification Test Results
Solubility | 633 g/l (20 °C) |
Melting Point | 89 - 90 °C |
Molar Mass | 68.08 g/mol |
Bulk Density | 500 - 600 kg/m3 |
Boiling Point | 256 °C (1013 hPa) |
Vapor Pressure | 0.003 hPa (20 °C) |
Flash Point | 145 °C |
Density | 1.030 g/cm3 |
pH | 10.5 (67 g/l, H2O, 20 °C) |
Ignition Point | 480 °C |
Hazard Class | 8 |
Hazard Class Description | Corrosive Material |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...