PvuII-HF®, Restriction Enzymes, New England Biolabs
Supplier: New England Biolabs (NEB)
R3151M
101641-160EA 294 USD
101641-160
PvuII-HF®, Restriction Enzymes, New England Biolabs
Nucleic Acid Reagents Restriction Enzymes
An E.coli strain that carries the cloned and modified PvuII gene from Proteus vulgaris (ATCC 13315).
High Fidelity (HF®) Restriction Enzymes have 100% activity in CutSmart® Buffer; single-buffer simplicity means more straightforward and streamlined sample processing. HF enzymes also exhibit dramatically reduced star activity. HF enzymes are all Time-Saver™ qualified and can therefore cut substrate DNA in 5-15 with the flexibility to digest overnight without degradation to DNA. Engineered with performance in mind, HF restriction enzymes are fully active under a broader range of conditions, minimizing off-target products, while offering flexibility in experimental design.