Aureothricin (from Streptomyces thioluteus) ≥98% (by HPLC)
Supplier: 0000027161
BVT-0345-C500
BVT-0345-M001
102513-786EA
55.38
USD
102513-786
102513-788
Aureothricin (from Streptomyces thioluteus) ≥98% (by HPLC)
Aureothricin
Aureothricin is a potent bacterial and yeast RNA polymerases inhibitor.
- Antibiotic
- Similar to thiolutin
- Anti-tumor compound
Active against Gram-positive and Gram-negative bacteria, yeast, filamentous fungi, protozoa and insects. Inhibitor of fungal mannan and glucan formation.
Yellow powder
After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.
Soluble in DMSO or dimethyl formamide; partially soluble in methanol or 100% ethanol; poorly soluble in water
Storage Temperature:
Freezer |
CAS Number:
574-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...