Reticulol (from Streptomyces species) ≥98% (by HPLC)

Supplier: 0000027161

BVT-0011-C250 BVT-0011-M001
102513-454EA 103.85 USD
102513-454 102513-456
Reticulol (from Streptomyces species) ≥98% (by HPLC)
Reticulol

Reticulol is a cyclic adenosine 3',5'-monophosphate phosphodiesterase and topoisomerase I (Topo I) inhibitor.


  • Antibiotic
  • Anti-tumor compound
  • Cyclic inhibitor


Antibiotic. Cyclic adenosine 3',5'-monophosphate phosphodiesterase and topoisomerase I (Topo I) inhibitor. Anti-tumor compound.


White to off-white solid


After reconstitution protect from light at -20°C. Stable for at least one year after receipt when stored at -20°C.


Soluble in DMSO, acetone or dimethylformamide.

Formula: C₁₁H₁₀O₅
Storage Temperature: Freezer
CAS Number: 26246-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR