GYY 4137 ≥95%

Supplier: 0000027161

AG-CR1-3513-M010 AG-CR1-3513-M050
102516-256EA 76.15 USD
102516-256 102516-258
GYY 4137 ≥95%
GYY 4137

Water-soluble, slow-releasing hydrogen sulfide (H2S) donor. Exhibits vasodilator and antihypertensive activity. Anti-inflammatory. Inhibits LPS-induced release of proinflammatory mediators (IL-1beta, IL-6, TNF-alpha, nitric oxide (NO) and PGE2) and increases the synthesis of the anti-inflammatory chemokine IL-10 through NF-kappaB, ATF-2 and HSP27 dependent pathways. Causes stomatal opening and reduces nitric oxide accumulation in plants. Oxidative stress-induced cell death inhibitor. Shows novel anti-cancer effects in vitro and in vivo. Anti-thrombotic via p-selectin dependent mechanism.

Formula: C₁₁H₁₆NO₂PS₂·C₄H₉NO
Storage Temperature: Freezer
CAS Number: 106740-09-4

Order Now

Specification Test Results

Purity 95% (NMR)
Appearance/Color White solid.
Solubility Soluble in DMSO or water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR