AK-7 ≥98%
Supplier: 0000027161
Synonyms:
3-(Azepan-1-ylsulphonyl)-N-(3-bromophenyl)benzamide, 3-(Azepan-1-ylsulfonyl)-N-(3-bromophenyl)benzamide
AG-CR1-3511-M005
AG-CR1-3511-M025
102516-248EA
110.77
USD
102516-248
102516-250
AK-7 ≥98%
AK-7
Brain-permeable SIRT2 (sirtuin 2) inhibitor.
Formula:
C₁₉H₂₁BrN₂O₃S Storage Temperature: Freezer |
MDL Number:
MFCD03140195 CAS Number: 420831-40-9 |
Specification Test Results
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...