α-NETA ≥97%
Supplier: 0000027161
|
Warning
|
AG-CR1-0151-M005
AG-CR1-0151-M025
102516-204EA
48.46
USD
102516-204
102516-206
α-NETA ≥97%
alpha-NETA
Potent fluorescent inhibitor of choline acetyltransferase. CMKLR1 antagonist. Inhibits chemerin-stimulated beta-arrestin2 association with CMKLR1, as well as chemerin-triggered CMKLR1+ cell migration. Aldehyde dehydrogenase 1 (ALDH1A1) antagonist.
Formula:
C₁₆H₂₀INO Storage Temperature: Freezer |
CAS Number:
31059-54-8 |
Specification Test Results
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in methanol or DMSO (3mg/ml). Sligthly soluble in water (1mg/ml). |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...