Bisindoylmaleimide X hydrochloride ≥98%

Supplier: 0000027161

Synonyms: Bisindolylmaleimide X HCl, Ro 31-8425, 3-(8-(Aminomethyl)-6,7,8,9-tetrahydropyrido[1,2-a]indol-10-yl)-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione hydrochloride

AG-CR1-0113-M001 AG-CR1-0113-M005
102516-166EA 96.92 USD
102516-166 102516-168
Bisindoylmaleimide X hydrochloride ≥98%
Bisindoylmaleimide X hydrochloride

Selective protein kinase C (PKC) inhibitor. Inhibits panel of protein kinases (e.g. GSK-3beta, CDK2). Inhibits superoxide generation in human neutrophils.

Formula: C₂₆H₂₄N₄O₂·HCl
Storage Temperature: Freezer
MDL Number: MFCD04972010
CAS Number: 145317-11-9

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color Red solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR