Ciglitazone ≥99%

Supplier: 0000027161
Warning

AG-CR1-0033-M001 AG-CR1-0033-M005 AG-CR1-0033-M025
102516-006EA 41.54 USD
102516-006 102516-008 102516-010
Ciglitazone ≥99%
Ciglitazone

Selective PPARgamma agonist (EC50=3 µM). Hypoglycemic agent. Angiogenesis inhibitor. Anti-inflammatory. Apoptosis inducer.

Formula: C₁₈H₂₃NO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD00865499
CAS Number: 74772-77-3

Order Now

Specification Test Results

Purity 99% (NMR)
Appearance/Color Off-white solid.
Solubility Soluble in DMSO or 100% ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR