1,5-Isoquinolinediol

Supplier: 0000027161
Warning

Synonyms: 5-Hydroxyisoquinoline-1(2H)-one

AG-CR1-0024-M005 AG-CR1-0024-M025 AG-CR1-0024-M100
102515-958EA 41.54 USD
102515-958 102515-960 102515-962
1,5-Isoquinolinediol
1,5-Isoquinolinediol

PARP-1 inhibitor. Neuroprotectant. Blocks nitric oxide-induced neuronal toxicity. Potent iNOS (inducible nitric oxide synthase/NOS II) inhibitor. Stimulates homologous recombination.

Formula: C₉H₇NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00006900
CAS Number: 5154-02-9

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO or methanol; insoluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR