MTEP ≥98%
Supplier: 0000027161
AG-CR1-0022-M005
AG-CR1-0022-M025
102515-950EA
96.92
USD
102515-950
102515-952
MTEP ≥98%
MTEP
Potent and selective antagonist for mGluR5. 5-fold higher anxiolytic activity than MPEP. Antidepressant. Neuroprotective. Produces antiparkinsonian-like effects in rats.
Formula:
C₁₁H₈N₂S Storage Temperature: Refrigerator |
CAS Number:
329205-68-7 |
Specification Test Results
Purity |
|
Appearance/Color | Light yellow solid. |
Solubility | Soluble in DMSO, 100% ethanol or methanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...