Ketanserin tartrate ≥98% (by NMR)

Supplier: 0000027161
Danger

Synonyms: 3-[2-[4-(4-Fluorobenzoyl)-1-piperdinyl)ethyl]-2,4(1H,3H)-quinazolinedione tartrate

AG-CR1-0014-M010 AG-CR1-0014-M050
102515-914EA 41.54 USD
102515-914 102515-916
Ketanserin tartrate ≥98% (by NMR)
Ketanserin tartrate

Selective 5-HT2A serotonin receptor antagonist. ATP-sensitive potassium channel inhibitor. hERG potassium channel blocker.

Formula: C₂₂H₂₂FN₃O₃·C₄H₆O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00084651
CAS Number: 83846-83-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purity >98% (NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in DMSO; slightly soluble in water (4 mg/ml) or 100% ethanol (3 mg/ml).


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR