Resveratrol
Supplier: 0000027161
|
Warning
|
Synonyms:
trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol
Potent phenolic antioxidant found in grapes and red wine. Eicosanoid synthesis and platelet aggregation inhibitor. Estrogen receptor agonist. Chemopreventive. Specific inhibitor of cyclooxygenase-1 (COX-1). Anti-inflammatory. Ribonucleotide reductase and DNA synthesis. Arrests cell cycle at S/G2 phase. Anticancer and antiproliferative compound. Apoptosis inducer. Protein kinase D inhibitor. Does not inhibit PKC. Autophagy inducer. Potent SIRT1 (sirtuin 1) activator. Neuroprotective. Adipogenesis inhibitor. PGC-1alpha activator. Sonic hedgehog (Shh) signaling pathway modulator. Gli1 mRNA expression inhibitor. Downregulates Gli transcriptional activity. Cardioprotective. Anti-diabetic. Senescence modulator. Extends lifespan.
Formula:
C₁₄H₁₂O₃ Storage Temperature: Freezer |
MDL Number:
MFCD00133799 CAS Number: 501-36-0 |
Specification Test Results
Purity |
|
Appearance/Color | Off-white powder. |
Solubility | Soluble in DMSO, ethanol or DMF. Slightly soluble in water or PBS. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...