Anti-NQO1 Mouse Monoclonal Antibody [clone: Skiny-1]

Supplier: Adipogen
AG-20A-0086-C050 AG-20A-0086-C100
102514-226EA 138.46 USD
102514-226 102514-228
Anti-NQO1 Mouse Monoclonal Antibody [clone: Skiny-1]
Antibodies
NQO1 is a cytosolic antioxidant flavoprotein that catalyzes the reduction of highly reactive quinone metabolites and their derivatives by using NAD(P)H as an electron donor. Thus NQO1 acts as a detoxyfying enzyme and is involved in the body's protection against oxidative stress. NQO1 acts as a protein chaperone, one of its targets being p53. In humans NQO1 is expressed at high levels in adipocytes and its expression levels are positively correlated with adiposity, glucose tolerance, and makers of liver dysfunction. Altered expression of NQO1 is associated with Alzheimer's disease. NQO1 is abnormally elevated in many types of solid tumors and may represent a useful biomarker of pancreatic cancer.

Type:
Antigen: NQO1
Clonality: Monoclonal
Clone: Skiny-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR