Anti-DLL1 Rat Monoclonal Antibody [clone: D1L357-1-4]

Supplier: Adipogen
AG-20A-0085-C050 AG-20A-0085-C100
102514-222EA 138.46 USD
102514-222 102514-224
Anti-DLL1 Rat Monoclonal Antibody [clone: D1L357-1-4]
Antibodies
The Notch ligand delta-like protein 1 (DLL1) is essential for postnatal arteriogenesis and contributes to tumor progression. DLL1 is involved in differentiation and self-renewal of adipocyte stem cells. Blocks the differentiation of progenitor cells into the B cell lineage while promoting the emergence of a population of cells with the characteristics of a T-cell/NK-cell precursor.

Type:
Antigen: DLL1
Clonality: Monoclonal
Clone: D1L357-1-4
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR