Anti-Calreticulin Mouse Monoclonal Antibody [clone: CR213-2AG]

Supplier: Adipogen
AG-20A-0079-C050 AG-20A-0079-C100
102514-198EA 249.23 USD
102514-198 102514-200
Anti-Calreticulin Mouse Monoclonal Antibody [clone: CR213-2AG]
Antibodies
Calreticulin is involved in regulation of intracellular Ca2+ homoeostasis and ER Ca2+ capacity. It constitutes together with calnexin and ERp57 the 'calreticulin/calnexin cycle' that is responsible for folding and quality control of newly synthesized glycoproteins. Calreticulin has been implicated to play a role in many biological systems, including functions inside and outside the ER, indicating that the protein is a multi-process molecule.

Type:
Antigen: Calreticulin
Clonality: Monoclonal
Clone: CR213-2AG
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR