Anti-FGF-23 Mouse Monoclonal Antibody [clone: FG322-3]

Supplier: Adipogen
AG-20A-0073-C050 AG-20A-0073-C100
102514-178EA 235.38 USD
102514-178 102514-180
Anti-FGF-23 Mouse Monoclonal Antibody [clone: FG322-3]
Antibodies
FGF-23 is a regulator of phosphate homeostasis. It upregulates EGR1 expression in the presence of KLBy. Acts directly on the parathyroid to decrease PTH secretion. Regulates the vitamin-D metabolism. Negatively regulates osteoblast differentiation and matrix mineralization. Defects in FGF-23 are the cause of autosomal dominant hypophosphataemic rickets (ADHR) and of hyperphosphatemic familial tumoral calcinosis (HFTC).

Type:
Antigen: FGF-23
Clonality: Monoclonal
Clone: FG322-3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR