Anti-IL-23p19 Rat Monoclonal Antibody [clone: I 178G]

Supplier: Adipogen
AG-20A-0027-C050 AG-20A-0027-C100
102514-012EA 138.46 USD
102514-012 102514-014
Anti-IL-23p19 Rat Monoclonal Antibody [clone: I 178G]
Antibodies
IL-23p19 associates with IL-12b to form the IL-23 interleukin, an heterodimeric cytokine which functions in innate and adaptive immunity. IL-23 binds to an heterodimeric receptor complex composed of IL-12RB1 and IL-23R, activates the Jak-Stat signaling cascade, stimulates memory rather than naive T-cells and promotes production of proinflammatory cytokines. IL-23 induces autoimmune inflammation and thus may be responsible for autoimmune inflammatory diseases and may be important for tumorigenesis.

Type:
Antigen: IL-23p19
Clonality: Monoclonal
Clone: I 178G
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR