Coumestrol ≥95% (by HPLC)

Supplier: Enzo Life Sciences
Warning

BML-S180-0005 BML-S180-0025
89162-578EA 122.08 USD
89162-578 89162-580
Coumestrol ≥95% (by HPLC)
Coumestrol

Estrogen receptor ligand.


  • Formulation: Light beige solid.


Phytoestrogen. Acts as an estrogen agonist.


Soluble in DMSO (25 mg/ml) and warm ethanol (5 mg/ml).

Formula: C₁₅H₈O₅
MW: 268.2 g/mol
Storage Temperature: Ambient
CAS Number: 479-13-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=488b7d3b7929aed0f4d9ed53bcfb9ae6a1ef96fb&vid=ffe916a6bf93d9e6cd58685ffabbeec4ff3cd814&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥95% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR