Anti-Proteasome 19S Rpn12/S14 Subunit Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW8815-0025 PW8815-0100
89162-222EA 203.83 USD
89162-222 89162-224
Anti-Proteasome 19S Rpn12/S14 Subunit Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human proteasome 26S (19S regulator complex) (Rpn12/S14 subunit).

Type: Primary
Antigen: PROTEASOME 19S RPN12/S14 SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR