Anti-Cop9 Signalosome Csn3 Subunit Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW8235-0025 PW8235-0100
89162-084EA 283.4 USD
89162-084 89162-086
Anti-Cop9 Signalosome Csn3 Subunit Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human COP9 signalosome (Csn3 subunit).

Type: Primary
Antigen: COP9 SIGNALOSOME CSN3 SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR