Anti-PSMA2 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW8105-0025 PW8105-0100
89161-984EA 171.13 USD
89161-984 89161-986
Anti-PSMA2 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, rabbit and cow proteasome 20S (alpha2 subunit).

Type: Primary
Antigen: PSMA2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR