Anti-SP100 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW0325-0025 PW0325-0100
89161-816EA 175.49 USD
89161-816 89161-818
Anti-SP100 Rabbit Polyclonal Antibody
Antibodies
Sp100 is an interferon-inducible protein and a major constituent of distinct nuclear domains, known as nuclear dots (NDs) or PML bodies. NDs play a role in autoimmunity, virus infections, and in the etiology of acute promyelocytic leukaemia. SUMO modification of Sp100 has been shown to promote interaction of Sp100 with non-histone chromosomal proteins, suggesting a regulatory role in chromatin organization and in functional interplay between these nuclear bodies and chromatin.

  • For Immunocytochemistry, Western Blot

Recognizes human Sp100 and SUMO modified Sp100 species.

Type: Primary
Antigen: SP100
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR