Ciglitazone ≥98%

Supplier: Enzo Life Sciences

BML-GR205-0005 BML-GR205-0025
89158-784EA 163.5 USD
89158-784 89158-786
Ciglitazone ≥98%
Ciglitazone

PPAR ligand


Ciglitazone is a thiazolidinedione type of antihyperglycemic agent. It displays antihyperglycemic activity in genetically obese C57 Bl/6 ob/ob mice and is a selective PPARgamma agonist (EC50=3 µM). Inhibits HUVEC differentiation and angiogenesis and also stimulates adipogenesis and decreases osteoblastogenesis in human mesenchymal stem cells.


Synthetic.

Formula: C₁₈H₂₃NO₃S
MDL Number: MFCD00865499
CAS Number: 74772-77-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d59f84e32eddd0d799ccaf5f90fd14d9fae62af0&vid=bd4c95fe5cdf7e038eadbddbcfbcf7492b01b0d3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR