Capsaicin ≥99% (by TLC)

Supplier: Enzo Life Sciences

BML-EI125-0200 BML-EI125-1000
89157-620EA 163.5 USD
89157-620 89157-622
Capsaicin ≥99% (by TLC)
Capsaicin

TRPV1 agonist


Capsaicin is an agonist at the TRPV1 receptor, a ligand-gated non-selective cation channel. Exhibits neuroprotective effects in a model of transient global cerebral ischemia. Induces release of substance P from afferent nociceptive specific neurons. Constituent of cayenne pepper. Powerful excitant of peropheral sensory nerve endings, specifically unmyelinated afferent neurons (C-fibers). Reversibly inhibits aggregation of platelets.


Isolated from Capsicum  fruit.

Formula: C₁₈H₂₇NO₃
MDL Number: MFCD00017259
CAS Number: 404-86-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a732662048f287c9a63bcc5659c6d1a7e8e2d4c&vid=5f959a4140b10bc046c5a172d1195d1d307382be&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR