Aflatoxin B2 ≥98% (by HPLC, TLC)
Supplier: Enzo Life Sciences
|
Danger
|
630-103-M001
630-103-M005
89152-992EA
163.5
USD
89152-992
89152-994
Aflatoxin B2 ≥98% (by HPLC, TLC)
Aflatoxin B2
Hepatotoxic and hepatocarcinogenic mycotoxin.
- Formulation: White to off-white powder; blue fluorescence.
Isolated from Aspergillus flavus.
Soluble in methanol, methylene chloride (5 mg/mL) or DMSO (5 mg/mL).
Synonym: MPL-A from E. coli, Serotype R515 (Re).
Formula:
C₁₇H₁₄O₆ MW: 314.3 g/mol Storage Temperature: Freezer |
CAS Number:
7220-81-7 EINECS: 230-618-8 UN: 3462 ADR: 6.1,I Merck Index: 14,00180 |
Specification Test Results
Purity | ≥98% (HPLC, TLC) |
Source | Isolated from Aspergillus flavus |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...