(2'Z,3'E)-6-Bromoindirubin-3'-oxime ≥98%
Supplier: Enzo Life Sciences
Synonyms:
GSK 3 Inhibitor IX, BIO, 6BIO, 6-Bromoindirubin-3'-oxime, (2Z,3E)-6'-Bromo-3-(hydroxyimino)-[2,3'-biindolinylidene]-2'-one
430-156-M001
430-156-M005
89151-272EA
98.1
USD
89151-272
76002-020
(2'Z,3'E)-6-Bromoindirubin-3'-oxime ≥98%
(2'Z,3'E)-6-Bromoindirubin-3'-oxime
Inibitor of phosphoinositide-dependent kinase 1 and GSK
Inhibitor of phosphoinositide-dependent kinase 1 (PDK1). Potent, reversible and ATP-competitive inhibitor of glycogen synthase kinase-3alpha/beta (GSK-3alpha/beta) (IC50=5nM).
Formula:
C₁₆H₁₀BrN₃O₂ MW: 356.18 g/mol |
MDL Number:
MFCD08705318 CAS Number: 667463-62-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...