Demecolcine ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: Colcemid

430-033-M001 430-033-M005
89150-938EA 55.59 USD
89150-938 89150-940
Demecolcine ≥98% (by HPLC)
Demecolcine

Depolymerizes microtubules and inhibits their formation. Induces apoptosis.


Semisynthetic from colchicine (Prod. No. ALX-380-033).

Formula: C₂₁H₂₅NO₅
MDL Number: MFCD00075459
CAS Number: 477-30-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b55f3f4412a851170582589b8bb9ffce3140bd&vid=45e5af23958fb5cb2884c65695975d0974f8d3fc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR