Myricetin ≥90% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 3,3',4',5,5',7-Hexahydroxyflavone, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-4H-chromen-4-one

385-012-M010 385-012-M050
89150-504EA 52.32 USD
89150-504 89150-506
Myricetin ≥90% (by HPLC)
Myricetin

Chemopreventive agent


Antioxidant flavonoid. Has antitumor and chemopreventive properties. Anti-inflammatory. Inhibits NF-kappaB activation.
PIM1 kinase inhibitor IC50=0.78µM.

Formula: C₁₅H₁₀O₈
MW: 318.24 g/mol
MDL Number: MFCD00006827
CAS Number: 529-44-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1511c042dc6035a4c34f0d7071cd4eb0b88e381a&vid=649d44dcb472c942c1e664f5bc73bebadf43b0e9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR