Polymyxin B sulphate
Supplier: Enzo Life Sciences
380-040-G001
380-040-G005
89149-876EA
101.37
USD
89149-876
89149-878
Polymyxin B sulphate
Polymyxin B sulphate
PKC inhibitor
- Mixture of Polymyxin B1, B2, B3 and B1-I
- Isolated from Bacillus polymyxa
- Antibiotic
Inhibitor of protein kinase C (PKC). Breaks the bacterial membrane by incorporating into the phospholipid of the outer membrane and activating phospholipase.
Formula:
C₅₅H₉₆N₁₆O₁₃·2H₂SO₄ MW: 1385.61 g/mol |
MDL Number:
MFCD00131991 CAS Number: 1405-20-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...