Celastrol ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: Tripterin

350-332-M005 350-332-M025
89149-518EA 114.45 USD
89149-518 89149-520
Celastrol ≥98% (by HPLC)
Celastrol

Anti-inflammatory and immunosuppressive


Cell permeable triterpenoid; anti-inflammatory and immunosuppressive compound. Antioxidant. Suppresses LPS-induced cytokine release in macrophages and monocytes (IC50=40nM for IL-1β and IL-1α, 80nM for IL-6, 110nM for prostaglandin E2 and 210nM for IL-8 and TNF-α). Suppresses nitric oxide (NO) production (IC50=230nM) and LPS-induced NF-κB activation (IC50=270nM). Inhibits chymotrypsin-like activity of 20S proteasome (IC50=2.5µM). Inhibits lipid peroxidation induced by ADP and Fe2+ in rat liver mitochondria (IC50=7µM). Induces expression of a wider set of heat shock proteins. Inhibits topoisomerase II.


Isolated from the root of Tripterygium sp.

Formula: C₂₉H₃₈O₄
MDL Number: MFCD03424073
CAS Number: 34157-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d7eb72f0f044ac7912af373881594d602b5ca83&vid=5342e4cceeb0fbfcd482b30933adc5c6e0ec9363&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR