D-Cycloserine

Supplier: BACHEM AMERICAS INC

Synonyms: D-(+)-Cycloserine, Cycloserine, D(+)-Cycloserine, (R)-4-Aminoisoxazolidin-3-one, D-4-Aminoisoxazolidin-3-one, (R)-(+)-Cycloserine

4030155.0005 4030155.0025
F-1480.0005BAEA 468.8 USD
F-1480.0005BA F-1480.0025BA
D-Cycloserine
D-Cycloserine

Sequence: D-Cycloserine
Synonym(s): (R)-4-Aminoisoxazalidin-3-one


Originally, D-cycloserine has been used as an antibiotic in the treatment of tuberculosis. More recently, it has found application in treating anxiety disorders (see the review by S.G.Hofmann et al.). Acting as a partial agonist of the NMDA receptor, the compound reduces neuropathic pain.

Formula: C₃H₆N₂O₂
MW: 102.09 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00005353
CAS Number: 68-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR