Human Transferrin (from Plasma), MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Transferrin is a glycoprotein with homologous N-terminal and C-terminal iron-binding domains.
- From Pooled Human Plasma
- Purity 99% by SDS-Page
- UV/Visible Absorbance 280 nm (water)
- Endotoxin: < 1 EU/mg By LAL
- Moisture Content: ≤ 5% By Karl Fisher
Transferrin is ideal for use as a growth factor for primary, continuous, and transformed cell cultures.
The normal human serum/plasma of each donor unit, from which this product was manufactured, has been tested and certified negative for the following by FDA approved testing procedures: HBsAg, Anti-HCV, Anti-HIV-1, Anti-HIV-2, Syphilis, ALT Test and NAT-PCR ( HAV, HIV,HBC,HCV and Parovirus B19)
Contents: 90% Iron Saturated; Contains no detectable IgG; Iron: 1350 μg/g By Diagnostic Assay
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...