Mouse Orexin B

Supplier: Thermo Scientific Chemicals

J65275-EXD J65275-MCR
AAJ65275-EXDEA 214 USD
AAJ65275-EXD AAJ65275-MCR
Mouse Orexin B
Proteins and Peptides

CAS No.: 202801-92-1
Molecular Formula: C126H214N45O34S
Formula Weight: 2935.38
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
Merck Reference: 14,6866
MDL No.: MFCD01635376

Order Now

SPECIFICATIONS

Protein/Peptide Name Orexin B
Environmentally Preferable
Species Mouse
Source
CAS No. 202801-92-1
MDL No. MFCD01635376
Color White to off-white
Form Solid
Molecular Weight 2935.38
Molecular Formula C₁₂₆H₂₁₄N₄₅O₃₄S
Storage Conditions -30 °C to -10 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR